Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRHL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195010.1 | NP_001181939.1 | 13 | 1652 | ||
NM_021180.3 | NP_067003.2 | 13 | 1548 | |||
NM_198173.2 | NP_937816.1 | 13 | 1717 | |||
NM_198174.2 | NP_937817.3 | 13 | 1717 | |||
XM_011541869.1 | XP_011540171.1 | 13 | 1598 | |||
XM_011541870.2 | XP_011540172.1 | 12 | 1519 | |||
GenBank mRNA | AK074386.1 | - | 13 | 1614 | ||
AK315164.1 | - | 13 | 1519 | |||
AL137763.1 | - | 1 | 33 | |||
AY231160.1 | - | 13 | 1548 | |||
AY231161.1 | - | 12 | 1361 | |||
BC036890.2 | - | 13 | 1663 | 5502632 |