Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RDH5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199771.1 | NP_001186700.1 | 2 | 480 | ||
NM_002905.3 | NP_002896.2 | 2 | 475 | |||
GenBank mRNA | AK293313.1 | - | 2 | 431 | ||
AK307157.1 | - | 2 | 302 | |||
BC028298.1 | - | 2 | 434 | 4942311 | ||
BI765016.1 | - | 2 | 386 | |||
BP381700.1 | - | 2 | 475 | |||
U43559.1 | - | 2 | 368 | |||
U89717.1 | - | 2 | 405 |