Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ROCK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005406.2 | NP_005397.1 | 28 | 4338 | ||
XM_011526137.2 | XP_011524439.1 | 17 | 2572 | |||
GenBank mRNA | BC113114.1 | - | 28 | 3413 | 7961989 | |
U43195.1 | - | 28 | 3397 |