Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP162 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286206.1 | NP_001273135.1 | 24 | 3805 | ||
NM_014895.3 | NP_055710.2 | 24 | 3859 | |||
XM_005248674.3 | XP_005248731.1 | 23 | 3982 | |||
XM_006715380.2 | XP_006715443.1 | 25 | 3953 | |||
XM_011535592.2 | XP_011533894.1 | 14 | 2713 | |||
XM_017010483.1 | XP_016865972.1 | 23 | 4538 | |||
XM_017010484.1 | XP_016865973.1 | 25 | 4156 | |||
GenBank mRNA | AB023226.1 | - | 24 | 3766 | ||
AK023613.1 | - | 15 | 2870 |