Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCTR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002980.2 | NP_002971.2 | 13 | 1739 | ||
XM_005263730.4 | XP_005263787.1 | 10 | 2193 | |||
XM_011511621.2 | XP_011509923.1 | 13 | 1754 | |||
XM_017004670.1 | XP_016860159.1 | 13 | 2205 | |||
XM_017004671.1 | XP_016860160.1 | 12 | 1646 | |||
XM_017004672.1 | XP_016860161.1 | 13 | 1569 | |||
XM_017004673.1 | XP_016860162.1 | 9 | 1112 | |||
GenBank mRNA | AK055551.1 | - | 10 | 2213 | ||
BC035757.1 | - | 13 | 1778 | 5763906 | ||
U13989.1 | - | 13 | 1563 | |||
U20178.1 | - | 13 | 1569 | |||
U28281.1 | - | 13 | 1621 |
Set Membership: |
G-protein coupled receptor |