Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBX6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004608.3 | NP_004599.2 | 5 | 851 | ||
XM_005255523.2 | XP_005255580.1 | 6 | 1136 | |||
XM_011545926.2 | XP_011544228.1 | 6 | 1299 | |||
XM_017023614.1 | XP_016879103.1 | 6 | 876 | |||
GenBank mRNA | AJ007989.1 | - | 5 | 815 | ||
AK022330.1 | - | 5 | 1485 | |||
BC026031.1 | - | 5 | 851 | 5460190 | ||
KJ534981.1 | - | 5 | 887 | |||
KJ535035.1 | - | 4 | 894 |