Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TLE3 SIRNA Assays ›
Gene Symbol:
TLE3
Gene Name:
transducin like enhancer of split 3
Gene Aliases:
ESG, ESG3, GRG3, HsT18976
Chromosome Location:
Chr.15: 70047790 - 70097935 on Build GRCh38
UniGene ID:
Hs.287362
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s224733
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TLE3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001105192.2
NP_001098662.1
10
1871
NM_001282979.1
NP_001269908.1
10
1871
NM_001282980.1
NP_001269909.1
10
1871
NM_001282981.1
NP_001269910.1
10
804
NM_001282982.1
NP_001269911.1
8
653
NM_005078.3
NP_005069.2
10
1871
NM_020908.2
NP_065959.1
10
1871
XM_005254622.4
XP_005254679.1
10
1919
XM_005254623.3
XP_005254680.1
10
1919
XM_005254625.3
XP_005254682.1
10
1919
XM_005254628.3
XP_005254685.1
10
1919
XM_005254633.2
XP_005254690.1
10
1889
XM_006720665.3
XP_006720728.1
10
1886
XM_011521976.2
XP_011520278.1
10
1919
XM_011521977.2
XP_011520279.1
10
1916
XM_011521978.2
XP_011520280.1
10
1919
XM_011521979.2
XP_011520281.1
9
725
XM_011521980.2
XP_011520282.1
10
1886
XM_011521981.1
XP_011520283.1
10
1919
XM_011521982.2
XP_011520284.1
8
1667
XM_011521983.2
XP_011520285.1
8
1667
XM_017022532.1
XP_016878021.1
10
1919
GenBank mRNA
AB046767.1
-
10
1853
AK096779.1
-
8
653
AK298482.1
-
10
774
AK315058.1
-
11
1199
AY583459.1
-
10
752
AY583460.1
-
10
752
BC041831.1
-
9
1068
5268249
BC043247.2
-
10
1870
5296274
M99438.1
-
10
773
Back To Top
More Information
Panther Classification:
Molecular Function -
transcription cofactor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
signal transduction
organ morphogenesis
Wnt signaling pathway
beta-catenin-TCF complex assembly
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-qqtbl:80/10.239.164.61:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0