Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF124 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297568.1 | NP_001284497.1 | 4 | 871 | ||
NM_001297569.1 | NP_001284498.1 | 4 | 675 | |||
NM_003431.3 | NP_003422.2 | 4 | 685 | |||
XM_005273257.4 | XP_005273314.1 | 4 | 738 | |||
XM_006711813.3 | XP_006711876.1 | 4 | 924 | |||
XM_011544272.2 | XP_011542574.1 | 4 | 722 | |||
GenBank mRNA | AK054647.1 | - | 4 | 684 | ||
BC096349.4 | - | 4 | 635 | 40009545 | ||
BC096350.4 | - | 4 | 635 | 40009546 | ||
BC096351.2 | - | 4 | 821 | 40009547 | ||
BC099661.3 | - | 4 | 821 | 40009541 | ||
DN993107.1 | - | 4 | 661 | |||
S54641.1 | - | 1 | 1076 |