Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290104.1 | NP_001277033.1 | 10 | 1427 | ||
NM_003460.2 | NP_003451.1 | 10 | 1427 | |||
XM_011545942.2 | XP_011544244.1 | 9 | 956 | |||
GenBank mRNA | AK293805.1 | - | 9 | 971 | ||
AK308820.1 | - | 9 | 971 | |||
AK310454.1 | - | 10 | 1427 | |||
AK313003.1 | - | 9 | 969 | |||
BC096304.3 | - | 9 | 944 | 40007464 | ||
BC096305.3 | - | 9 | 944 | 40007468 | ||
BC096306.1 | - | 9 | 944 | 40007471 | ||
BC096307.3 | - | 9 | 944 | 40007473 | ||
M90366.1 | - | 9 | 944 |