Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF212 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012256.3 | NP_036388.2 | 4 | 704 | ||
XM_005250055.2 | XP_005250112.2 | 4 | 702 | |||
XM_011516581.2 | XP_011514883.2 | 4 | 692 | |||
XM_011516582.1 | XP_011514884.1 | 4 | 624 | |||
XM_017012646.1 | XP_016868135.1 | 4 | 705 | |||
XM_017012647.1 | XP_016868136.1 | 5 | 658 | |||
XM_017012648.1 | XP_016868137.1 | 4 | 947 | |||
GenBank mRNA | AK074821.1 | - | 5 | 707 | ||
AK315086.1 | - | 4 | 713 | |||
BC022785.2 | - | 4 | 671 | 3849803 | ||
U38864.1 | - | 2 | 155 |