Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCUBE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173050.3 | NP_766638.2 | 14 | 1787 | ||
XM_005261752.1 | XP_005261809.1 | 15 | 1838 | |||
XM_011530389.2 | XP_011528691.1 | 15 | 1832 | |||
XM_011530390.2 | XP_011528692.1 | 14 | 1717 | |||
GenBank mRNA | AF525689.1 | - | 15 | 1680 | ||
AK127719.1 | - | 11 | 1260 |