Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: STAM | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001324283.1 | NP_001311212.1 | 2, 3 | 330 | ||
NM_001324284.1 | NP_001311213.1 | 4, 5 | 519 | |||
NM_001324285.1 | NP_001311214.1 | 4, 5 | 527 | |||
NM_001324288.1 | NP_001311217.1 | 3, 4 | 415 | |||
NM_001324289.1 | NP_001311218.1 | 2, 3 | 330 | |||
NM_003473.3 | NP_003464.1 | 3, 4 | 415 | |||
XM_011519695.2 | XP_011517997.1 | 3, 4 | 391 | |||
GenBank mRNA | AK314405.1 | - | 3, 4 | 299 | ||
AU118708.1 | - | 3, 4 | 416 | |||
BC030586.2 | - | 3, 4 | 358 | 4820857 | ||
U43899.1 | - | 3, 4 | 242 |