Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PUDP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135565.1 | NP_001129037.1 | 3 | 349 | ||
NM_001178135.1 | NP_001171606.1 | 2 | 280 | |||
NM_012080.4 | NP_036212.3 | 2 | 280 | |||
GenBank mRNA | AA262524.1 | - | 1 | 157 | ||
AK223405.1 | - | 2 | 238 | |||
AK300740.1 | - | 2 | 215 | |||
AK307088.1 | - | 2 | 235 | |||
AK313155.1 | - | 2 | 243 | |||
AU099056.1 | - | 2 | 238 | |||
AU141098.2 | - | 2 | 272 | |||
BC012494.1 | - | 2 | 215 | 4478358 | ||
BG419948.1 | - | 224 | ||||
DA653367.1 | - | 2 | 230 | |||
DC388394.1 | - | 2 | 281 | |||
DR156836.1 | - | 3 | 317 | |||
M86934.1 | - | 2 | 196 |