Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289111.1 | NP_001276040.1 | 9 | 818 | ||
NM_032194.2 | NP_115570.1 | 10 | 856 | |||
XM_017011354.1 | XP_016866843.1 | 7 | 1292 | |||
GenBank mRNA | AK292165.1 | - | 10 | 835 | ||
AL833613.1 | - | 9 | 786 | |||
BC035314.1 | - | 10 | 785 | 4745524 |