Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOXL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289164.1 | NP_001276093.1 | 9 | 1576 | ||
NM_001289165.1 | NP_001276094.1 | 7 | 1293 | |||
NM_032603.3 | NP_115992.1 | 11 | 2011 | |||
XM_011533134.2 | XP_011531436.1 | 11 | 1971 | |||
XM_017005112.1 | XP_016860601.1 | 9 | 1615 | |||
GenBank mRNA | AF282619.1 | - | 11 | 1984 | ||
AF284815.1 | - | 11 | 1991 | |||
AF311313.1 | - | 11 | 1982 | |||
AK309474.1 | - | 9 | 1573 | |||
AK309478.1 | - | 9 | 2192 | |||
BC033130.1 | - | 4 | 517 | |||
BC071865.1 | - | 9 | 1600 | 6650423 | ||
DQ378059.1 | - | 7 | 1293 |