Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBRG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032811.2 | NP_116200.2 | 5, 6 | 978 | ||
NR_016021.1 | 4, 5 | 841 | ||||
XM_005271709.3 | XP_005271766.1 | 2, 3 | 784 | |||
XM_017018442.1 | XP_016873931.1 | 2, 3 | 1799 | |||
GenBank mRNA | AK027527.1 | - | 4, 5 | 812 | ||
AK074140.1 | - | 4, 5 | 565 | |||
AK222936.1 | - | 4, 5 | 817 | |||
AL713631.1 | - | 1, 2 | 83 | |||
AY696294.1 | - | 3, 4 | 346 | |||
BC041043.1 | - | 4, 5 | 826 | |||
BC064392.1 | - | 4, 5 | 790 | |||
BC107428.1 | - | 4, 5 | 831 | |||
BC109269.2 | - | 3, 4 | 460 | |||
DQ144542.1 | - | 5, 6 | 740 |