Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGS5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001254748.1 | NP_001241677.1 | 3 | 281 | ||
NM_001254749.1 | NP_001241678.1 | 2 | 396 | |||
NM_003617.3 | NP_003608.1 | 2 | 396 | |||
GenBank mRNA | AB008109.1 | - | 2 | 139 | ||
AF030108.1 | - | 2 | 134 | |||
AF159570.1 | - | 2 | 102 | |||
AF493929.1 | - | 2 | 58 | |||
AK315357.1 | - | 2 | 139 | |||
AU130764.1 | - | 2 | 305 | |||
BC030059.1 | - | 2 | 121 | 4098248 | ||
BP292867.1 | - | 3 | 241 | |||
BU172751.1 | - | 2 | 126 | |||
BX537427.1 | - | 2 | 140 | |||
DA699380.1 | - | 2 | 374 | |||
DA731601.1 | - | 2 | 397 |