Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other KYAT1 SIRNA Assays ›
Gene Symbol:
KYAT1
Gene Name:
kynurenine aminotransferase 1
Gene Aliases:
CCBL1, GTK, KAT1, KATI
Chromosome Location:
Chr.9: 128832942 - 128882551 on Build GRCh38
UniGene ID:
Hs.495250
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s225044
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: KYAT1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001122671.1
NP_001116143.1
3
357
NM_001122672.1
NP_001116144.1
3
363
NM_001287390.1
NP_001274319.1
5
754
NM_004059.4
NP_004050.3
3
363
NR_109829.1
3
363
XM_006717311.3
XP_006717374.1
4
549
XM_011519165.2
XP_011517467.1
5
725
XM_011519167.2
XP_011517469.2
3
329
XM_011519168.1
XP_011517470.1
2
188
XM_011519169.1
XP_011517471.1
3
457
XM_011519170.2
XP_011517472.1
3
825
XM_017015260.1
XP_016870749.1
4
509
XM_017015261.1
XP_016870750.1
5
731
XM_017015262.1
XP_016870751.1
5
1220
XM_017015263.1
XP_016870752.1
5
635
XM_017015264.1
XP_016870753.1
4
542
XM_017015265.1
XP_016870754.1
3
919
XM_017015266.1
XP_016870755.1
4
550
XM_017015267.1
XP_016870756.1
3
371
XM_017015268.1
XP_016870757.1
4
533
GenBank mRNA
AK297995.1
-
5
610
AK314427.1
-
3
233
AU123740.1
-
3
341
BC021262.2
-
3
324
4137942
BC033685.1
-
3
217
5193675
BI755163.1
-
3
217
DA381449.1
-
3
363
EL733697.1
-
2
178
X82224.1
-
2
178
Back To Top
More Information
Panther Classification:
Molecular Function -
transaminase
Gene Ontology Categories:
Function(s)
Process(es)
L-phenylalanine catabolic process
tryptophan catabolic process
cellular modified amino acid metabolic process
cellular amino acid biosynthetic process
kynurenine metabolic process
L-kynurenine catabolic process
protein binding
transaminase activity
kynurenine-oxoglutarate transaminase activity
pyridoxal phosphate binding
protein homodimerization activity
L-phenylalanine:pyruvate aminotransferase activity
glutamine-phenylpyruvate transaminase activity
cysteine-S-conjugate beta-lyase activity
L-glutamine:pyruvate aminotransferase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-7cf5v:80/10.239.163.60:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0