Gene Symbol: MBD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003927.4 | NP_003918.1 | 1 | 755 | ||
NM_015832.4 | NP_056647.1 | 1 | 755 | |||
GenBank mRNA | AF072242.1 | - | 1 | 755 | ||
AF072246.1 | - | 1 | 58 | |||
BC032638.1 | - | 1 | 596 | 5496721 | ||
BQ228712.1 | - | 474 |