Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UNC119 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005148.3 | NP_005139.1 | 5 | 1190 | ||
NM_054035.2 | NP_473376.1 | 4 | 1457 | |||
XM_017025289.1 | XP_016880778.1 | 6 | 1215 | |||
GenBank mRNA | AB593014.1 | - | 4 | 1460 | ||
AF028788.1 | - | 5 | 1122 | |||
AF028789.1 | - | 4 | 1390 | |||
AK124531.1 | - | 4 | 3996 | |||
BC027176.1 | - | 5 | 1164 | 4341426 | ||
BX641031.1 | - | 4 | 3919 | |||
BX647747.1 | - | 2 | 1022 | |||
BX648530.1 | - | 3 | 1860 | |||
U40998.1 | - | 5 | 1173 |