Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014207.3 | NP_055022.2 | 9 | 1451 | ||
GenBank mRNA | AK292698.1 | - | 9 | 1420 | ||
BC027901.1 | - | 9 | 1414 | 5228913 | ||
X04391.1 | - | 9 | 1420 |