Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHSY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014918.4 | NP_055733.2 | 3 | 2565 | ||
XM_006720435.3 | XP_006720498.1 | 3 | 1847 | |||
XM_011521364.2 | XP_011519666.1 | 4 | 2188 | |||
XM_017022011.1 | XP_016877500.1 | 2 | 1621 | |||
GenBank mRNA | AB023207.1 | - | 3 | 2583 | ||
AB071402.1 | - | 3 | 2103 | |||
AK075096.1 | - | 1 | 295 | |||
AK296943.1 | - | 2 | 1342 | |||
AY358529.1 | - | 3 | 2577 | |||
BC046247.1 | - | 3 | 2572 | 5751618 |