Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NLRP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033053.2 | NP_001028225.1 | 12 | 3940 | ||
NM_014922.4 | NP_055737.1 | 12 | 3928 | |||
NM_033004.3 | NP_127497.1 | 12 | 3928 | |||
NM_033006.3 | NP_127499.1 | 11 | 3838 | |||
NM_033007.3 | NP_127500.1 | 11 | 3838 | |||
GenBank mRNA | AB023143.1 | - | 12 | 3895 | ||
AF229059.1 | - | 12 | 3373 | |||
AF229060.1 | - | 12 | 3373 | |||
AF229061.1 | - | 11 | 3283 | |||
AF229062.1 | - | 11 | 3283 | |||
AF298548.1 | - | 13 | 3532 | |||
AF310105.1 | - | 12 | 3891 | |||
AK026393.1 | - | 9 | 1851 | |||
AK026398.1 | - | 9 | 1201 | |||
AK310936.1 | - | 13 | 3739 | |||
AL117470.1 | - | 9 | 2531 |