Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: FNDC3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079673.1 | NP_001073141.1 | 23, 24 | 3296 | ||
NM_001278438.1 | NP_001265367.1 | 23, 24 | 3339 | |||
NM_014923.4 | NP_055738.3 | 21, 22 | 2920 | |||
NR_103528.1 | 23, 24 | 3328 | ||||
XM_011534997.2 | XP_011533299.1 | 22, 23 | 3010 | |||
XM_017020440.1 | XP_016875929.1 | 23, 24 | 3181 | |||
XM_017020441.1 | XP_016875930.1 | 25, 26 | 3802 | |||
XM_017020442.1 | XP_016875931.1 | 24, 25 | 3334 | |||
GenBank mRNA | AB023187.2 | - | 21, 22 | 2850 | ||
AJ749706.1 | - | 23, 24 | 3298 | |||
AJ749707.1 | - | 21, 22 | 2911 | |||
AK172814.1 | - | 1, 2 | 227 | |||
AK302415.1 | - | 21, 22 | 2920 | |||
BC060816.1 | - | 14, 15 | 1958 | |||
BC070072.1 | - | 14, 15 | 1958 | |||
BC132812.1 | - | 23, 24 | 3063 | 40146834 | ||
BC136617.1 | - | 23, 24 | 3064 | 9020607 | ||
BC144301.1 | - | 23, 24 | 3064 | 9052821 | ||
BX640739.1 | - | 23, 24 | 3199 | |||
BX648141.1 | - | 23, 24 | 3286 | |||
BX648706.1 | - | 25, 26 | 3144 |