Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CNKSR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168647.1 | NP_001162118.1 | 20 | 3114 | ||
NM_014927.3 | NP_055742.2 | 21 | 3204 | |||
GenBank mRNA | AF418269.1 | - | 21 | 2760 | ||
BC126121.1 | - | 21 | 2759 | 8991837 | ||
BC136289.1 | - | 21 | 2759 | 9020276 | ||
BC143839.1 | - | 20 | 2669 | 9052354 |