Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other C1ORF61 SIRNA Assays ›
Gene Symbol:
C1orf61
Gene Name:
chromosome 1 open reading frame 61
Gene Aliases:
CROC4
Chromosome Location:
Chr.1: 156404252 - 156456649 on Build GRCh38
UniGene ID:
Hs.380027
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s225438
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: C1orf61
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001320453.1
NP_001307382.1
5
644
NM_001320454.1
NP_001307383.1
4
646
NM_001320455.1
NP_001307384.1
4
597
NM_006365.2
NP_006356.1
7
896
NR_135260.1
4
548
NR_135261.1
5
633
NR_135262.1
3
448
NR_135263.1
5
682
NR_135264.1
6
800
NR_135265.1
2
412
NR_135266.1
3
485
NR_135267.1
3
512
NR_135268.1
7
1130
XM_011509067.1
XP_011507369.1
6
795
XM_011509068.1
XP_011507370.1
5
695
XM_011509070.1
XP_011507372.1
6
1209
XM_011509071.2
XP_011507373.1
6
1996
XM_011509072.1
XP_011507374.1
6
819
XM_011509073.2
XP_011507375.1
5
983
XM_011509074.1
XP_011507376.1
5
764
XM_017000062.1
XP_016855551.1
10
1377
XM_017000063.1
XP_016855552.1
8
1001
XM_017000064.1
XP_016855553.1
6
918
XM_017000065.1
XP_016855554.1
8
1068
XM_017000066.1
XP_016855555.1
8
1125
GenBank mRNA
AK091014.1
-
1
2431
AK094493.1
-
1
1896
AK124566.1
-
1
4628
AK124870.1
-
1
3452
BC016978.1
-
1
1050
BC050448.1
-
5
547
BC050741.1
-
3
339
BC054045.1
-
4
579
BC059943.1
-
5
533
BC107686.1
-
4
407
BF347271.1
-
581
BG716287.1
-
5
524
BI597719.1
-
3
331
BI668594.1
-
5
566
DB534652.2
-
4
575
DN990706.1
-
4
494
HY121703.1
-
2
413
U49857.1
-
7
740
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0