Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RABAC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006423.2 | NP_006414.2 | 2 | 298 | ||
GenBank mRNA | AF112202.1 | - | 2 | 228 | ||
AF112996.1 | - | 2 | 224 | |||
AJ133534.1 | - | 2 | 244 | |||
BC008950.2 | - | 2 | 250 | 2963628 | ||
BT019964.1 | - | 2 | 214 | |||
CF242972.1 | - | 2 | 253 | |||
CR457214.1 | - | 2 | 214 | |||
CR542116.1 | - | 2 | 214 | |||
DB454056.1 | - | 2 | 298 |