Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DIDO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193369.1 | NP_001180298.1 | 4 | 1295 | ||
NM_001193370.1 | NP_001180299.1 | 4 | 1295 | |||
NM_022105.4 | NP_071388.2 | 4 | 1339 | |||
NM_033081.2 | NP_149072.2 | 4 | 1339 | |||
NM_080796.3 | NP_542986.1 | 4 | 1302 | |||
NM_080797.3 | NP_542987.2 | 4 | 1339 | |||
XM_011528504.1 | XP_011526806.1 | 4 | 1272 | |||
XM_011528505.1 | XP_011526807.1 | 4 | 1339 | |||
XM_011528506.1 | XP_011526808.1 | 4 | 8123 | |||
XM_011528507.1 | XP_011526809.1 | 4 | 1207 | |||
XM_011528508.1 | XP_011526810.1 | 4 | 1348 | |||
XM_011528509.2 | XP_011526811.1 | 4 | 1272 | |||
GenBank mRNA | AB002331.2 | - | 4 | 1271 | ||
AK002127.1 | - | 4 | 1283 | |||
AY481571.1 | - | 4 | 1247 | |||
AY481572.1 | - | 4 | 1247 | |||
BC000770.2 | - | 3 | 1183 | |||
BC004237.2 | - | 5 | 1264 | |||
BC014489.1 | - | 4 | 1220 | |||
BC137177.1 | - | 3 | 1194 | 9021174 |