Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138409.2 | NP_612418.2 | 3, 4 | 423 | ||
XM_011535400.2 | XP_011533702.1 | 3, 4 | 349 | |||
XM_017010219.1 | XP_016865708.1 | 4, 5 | 721 | |||
XM_017010220.1 | XP_016865709.1 | 3, 4 | 990 | |||
GenBank mRNA | AK090775.1 | - | 3, 4 | 423 | ||
AK292736.1 | - | 3, 4 | 354 | |||
BC010003.2 | - | 3, 4 | 368 | 4134852 | ||
BC039855.1 | - | 3, 4 | 286 | 3850270 |