Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308174.1 | NP_001295103.1 | 10 | 1514 | ||
NM_138790.3 | NP_620145.2 | 10 | 1476 | |||
XM_011536411.2 | XP_011534713.1 | 10 | 1524 | |||
XM_017020965.1 | XP_016876454.1 | 10 | 1413 | |||
GenBank mRNA | AK295352.1 | - | 10 | 1469 | ||
AK296188.1 | - | 10 | 1514 | |||
AY358843.1 | - | 10 | 1442 | |||
BC015003.1 | - | 10 | 1405 | 4853026 |