Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MFSD12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287529.1 | NP_001274458.1 | 5 | 1030 | ||
NM_174983.4 | NP_778148.2 | 5 | 1057 | |||
XM_005259490.4 | XP_005259547.1 | 5 | 1065 | |||
XM_006722647.3 | XP_006722710.1 | 5 | 1064 | |||
XM_011527684.2 | XP_011525986.1 | 5 | 1063 | |||
XM_017026288.1 | XP_016881777.1 | 5 | 1064 | |||
GenBank mRNA | BC036706.2 | - | 5 | 1009 | 4623094 | |
BC068439.1 | - | 5 | 1046 | 5314853 |