Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYPD6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195685.1 | NP_001182614.1 | 3 | 485 | ||
NM_194317.3 | NP_919298.1 | 3 | 475 | |||
GenBank mRNA | AK124733.1 | - | 3 | 485 | ||
AK309248.1 | - | 3 | 327 | |||
AY358623.1 | - | 3 | 391 | |||
BC032836.1 | - | 5 | 695 | |||
BC047013.1 | - | 4 | 505 | 5171270 | ||
DA246384.1 | - | 3 | 476 |