Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178003.1 | NP_001171474.1 | 5 | 1148 | ||
NM_144668.5 | NP_653269.3 | 5 | 1135 | |||
GenBank mRNA | AA853421.1 | - | 2 | 160 | ||
AL833930.1 | - | 3 | 423 | |||
BC028421.2 | - | 5 | 1133 | 4826893 | ||
BC036233.1 | - | 5 | 1128 | 5295406 |