Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001178003.1 | NP_001171474.1 | 8 | 1421 | ||
NM_144668.5 | NP_653269.3 | 8 | 1408 | |||
GenBank mRNA | AL833930.1 | - | 6 | 696 | ||
BC028421.2 | - | 8 | 1406 | 4826893 | ||
BC036233.1 | - | 8 | 1401 | 5295406 |