Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKRD46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270377.1 | NP_001257306.1 | 3 | 475 | ||
NM_001270378.1 | NP_001257307.1 | 3 | 1073 | |||
NM_001270379.1 | NP_001257308.1 | 3 | 475 | |||
NM_198401.3 | NP_940683.1 | 4 | 580 | |||
XM_006716509.1 | XP_006716572.1 | 3 | 616 | |||
XM_017013106.1 | XP_016868595.1 | 3 | 538 | |||
GenBank mRNA | AB077205.1 | - | 4 | 578 | ||
AK095235.1 | - | 3 | 450 | |||
AL832842.1 | - | 3 | 435 | |||
BC050396.1 | - | 3 | 431 | 6065488 | ||
BC060843.1 | - | 3 | 448 | 30341860 | ||
DA503253.1 | - | 3 | 475 |