Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ANKRD6 SIRNA Assays ›
Gene Symbol:
ANKRD6
Gene Name:
ankyrin repeat domain 6
Gene Aliases:
-
Chromosome Location:
Chr.6: 89433178 - 89633834 on Build GRCh38
UniGene ID:
Hs.702213
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s22577
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ANKRD6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242809.1
NP_001229738.1
13
1658
NM_001242811.1
NP_001229740.1
13
2041
NM_001242813.1
NP_001229742.1
12
1553
NM_001242814.1
NP_001229743.1
10
1279
NM_014942.4
NP_055757.3
13
1658
XM_005248679.3
XP_005248736.1
12
1526
XM_005248680.3
XP_005248737.1
14
3830
XM_005248681.2
XP_005248738.1
13
1759
XM_005248682.3
XP_005248739.1
11
1448
XM_005248683.3
XP_005248740.1
10
1328
XM_005248684.3
XP_005248741.1
7
1046
XM_011535596.2
XP_011533898.1
11
1427
XM_011535597.2
XP_011533899.1
11
1427
XM_011535598.2
XP_011533900.1
12
1528
XM_011535599.2
XP_011533901.1
12
1528
XM_011535600.2
XP_011533902.1
11
1450
XM_017010493.1
XP_016865982.1
11
1421
XM_017010494.1
XP_016865983.1
11
1427
XM_017010495.1
XP_016865984.1
12
1528
XM_017010496.1
XP_016865985.1
12
1692
XM_017010497.1
XP_016865986.1
11
1361
XM_017010498.1
XP_016865987.1
12
1522
XM_017010499.1
XP_016865988.1
10
1322
XM_017010500.1
XP_016865989.1
10
1328
XM_017010501.1
XP_016865990.1
11
1429
XM_017010502.1
XP_016865991.1
11
3110
XM_017010503.1
XP_016865992.1
11
3114
XM_017010504.1
XP_016865993.1
10
1256
XM_017010505.1
XP_016865994.1
11
1423
XM_017010506.1
XP_016865995.1
10
1262
XM_017010507.1
XP_016865996.1
10
1262
XM_017010508.1
XP_016865997.1
10
3006
XM_017010509.1
XP_016865998.1
9
1157
XM_017010510.1
XP_016865999.1
9
1212
XM_017010511.1
XP_016866000.1
9
2915
XM_017010512.1
XP_016866001.1
8
1147
XM_017010513.1
XP_016866002.1
8
1279
XM_017010514.1
XP_016866003.1
7
1048
XM_017010515.1
XP_016866004.1
13
1543
GenBank mRNA
AB023174.1
-
12
1380
AK074189.1
-
4
581
AK092235.1
-
4
1233
AK096867.1
-
10
1141
AK295731.1
-
13
1958
AK307792.1
-
13
1627
AL117504.1
-
10
1059
BC001078.2
-
1
95
BC042173.1
-
13
1623
5454509
Back To Top
More Information
Panther Classification:
Molecular Function -
protein-binding activity modulator
kinase inhibitor
kinase modulator
non-receptor serine/threonine protein kinase
ubiquitin-protein ligase
protein modifying enzyme
Gene Ontology Categories:
Function(s)
positive regulation of JNK cascade
negative regulation of canonical Wnt signaling pathway
positive regulation of Wnt signaling pathway, planar cell polarity pathway
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-qqtbl:80/10.239.164.61:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0