Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CLSTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009566.2 | NP_001009566.1 | 10 | 2178 | ||
NM_001302883.1 | NP_001289812.1 | 10 | 2178 | |||
NM_014944.4 | NP_055759.3 | 9 | 2148 | |||
GenBank mRNA | AB020718.1 | - | 10 | 2178 | ||
AF438482.1 | - | 9 | 2114 | |||
AK000605.1 | - | 6 | 845 | |||
AK289798.1 | - | 9 | 1356 | |||
AK304813.1 | - | 10 | 1420 | |||
AY753301.1 | - | 9 | 1355 | |||
BC033902.2 | - | 9 | 1606 | 5263192 |