Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC125 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297696.1 | NP_001284625.1 | 11 | 1215 | ||
NM_001297697.1 | NP_001284626.1 | 13 | 1271 | |||
NM_176816.4 | NP_789786.2 | 12 | 1432 | |||
XM_005248458.4 | XP_005248515.1 | 12 | 1669 | |||
XM_005248459.4 | XP_005248516.1 | 12 | 1471 | |||
XM_005248461.3 | XP_005248518.1 | 11 | 1158 | |||
XM_005248463.3 | XP_005248520.1 | 10 | 1026 | |||
XM_005248464.3 | XP_005248521.1 | 11 | 1299 | |||
XM_006714569.3 | XP_006714632.1 | 12 | 1459 | |||
XM_006714570.3 | XP_006714633.1 | 12 | 1243 | |||
XM_011543255.2 | XP_011541557.1 | 12 | 1449 | |||
XM_011543256.2 | XP_011541558.1 | 12 | 1450 | |||
XM_011543257.2 | XP_011541559.1 | 11 | 1366 | |||
XM_011543258.2 | XP_011541560.1 | 12 | 1474 | |||
XM_011543259.2 | XP_011541561.1 | 12 | 1474 | |||
XM_017009206.1 | XP_016864695.1 | 12 | 1664 | |||
XM_017009207.1 | XP_016864696.1 | 12 | 1459 | |||
XM_017009208.1 | XP_016864697.1 | 10 | 1298 | |||
XM_017009209.1 | XP_016864698.1 | 11 | 1296 | |||
XM_017009210.1 | XP_016864699.1 | 9 | 1219 | |||
XM_017009211.1 | XP_016864700.1 | 6 | 701 | |||
GenBank mRNA | AB024691.1 | - | 12 | 1432 | ||
AB024692.1 | - | 11 | 1135 | |||
AK299871.1 | - | 13 | 1271 | |||
BC113749.1 | - | 6 | 684 |