Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC125 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297697.1 | NP_001284626.1 | 11 | 921 | ||
NM_176816.4 | NP_789786.2 | 10 | 1082 | |||
XM_005248458.4 | XP_005248515.1 | 10 | 1319 | |||
XM_005248459.4 | XP_005248516.1 | 10 | 1121 | |||
XM_005248461.3 | XP_005248518.1 | 9 | 808 | |||
XM_005248463.3 | XP_005248520.1 | 8 | 676 | |||
XM_006714569.3 | XP_006714632.1 | 10 | 1109 | |||
XM_006714570.3 | XP_006714633.1 | 10 | 893 | |||
XM_011543255.2 | XP_011541557.1 | 10 | 1099 | |||
XM_011543256.2 | XP_011541558.1 | 10 | 1100 | |||
XM_011543257.2 | XP_011541559.1 | 9 | 1016 | |||
XM_011543258.2 | XP_011541560.1 | 10 | 1124 | |||
XM_011543259.2 | XP_011541561.1 | 10 | 1124 | |||
XM_017009206.1 | XP_016864695.1 | 10 | 1314 | |||
XM_017009207.1 | XP_016864696.1 | 10 | 1109 | |||
XM_017009208.1 | XP_016864697.1 | 8 | 948 | |||
XM_017009210.1 | XP_016864699.1 | 7 | 869 | |||
XM_017009211.1 | XP_016864700.1 | 4 | 351 | |||
GenBank mRNA | AB024691.1 | - | 10 | 1082 | ||
AB024692.1 | - | 9 | 785 | |||
AK299871.1 | - | 11 | 921 |