Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLAC9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012973.1 | NP_001012991.1 | 2 | 404 | ||
GenBank mRNA | BC066348.1 | - | 2 | 404 | 30339526 | |
BC090922.1 | - | 2 | 404 | 30331098 | ||
BC117332.1 | - | 2 | 232 | 40125883 | ||
BC117334.1 | - | 2 | 232 | 40125885 |