Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEP152 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001194998.1 | NP_001181927.1 | 17 | 2353 | ||
NM_014985.3 | NP_055800.2 | 17 | 2353 | |||
XM_006720437.3 | XP_006720500.1 | 17 | 2538 | |||
XM_011521373.2 | XP_011519675.1 | 17 | 3106 | |||
XM_011521374.2 | XP_011519676.1 | 17 | 3105 | |||
XM_011521375.2 | XP_011519677.1 | 17 | 3105 | |||
XM_011521378.2 | XP_011519680.1 | 17 | 3105 | |||
XM_011521379.2 | XP_011519681.1 | 17 | 3104 | |||
XM_011521381.2 | XP_011519683.1 | 5 | 1266 | |||
XM_017022014.1 | XP_016877503.1 | 10 | 1299 | |||
XM_017022015.1 | XP_016877504.1 | 5 | 455 | |||
XM_017022016.1 | XP_016877505.1 | 17 | 3108 | |||
GenBank mRNA | AB020719.1 | - | 16 | 1958 | ||
AK303805.1 | - | 17 | 2276 | |||
BC117182.1 | - | 17 | 2194 | 40125733 | ||
BX648822.1 | - | 17 | 2280 |