Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other EHBP1 SIRNA Assays ›
Gene Symbol:
EHBP1
Gene Name:
EH domain binding protein 1
Gene Aliases:
HPC12, NACSIN
Chromosome Location:
Chr.2: 62673851 - 63046487 on Build GRCh38
UniGene ID:
Hs.271667
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s225945
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: EHBP1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001142614.1
NP_001136086.1
10
1572
NM_001142615.2
NP_001136087.1
11
1805
NM_001142616.1
NP_001136088.1
11
1759
NM_015252.3
NP_056067.2
12
1864
XM_005264225.2
XP_005264282.1
12
1762
XM_005264226.2
XP_005264283.1
12
2119
XM_005264227.1
XP_005264284.1
11
1718
XM_005264228.2
XP_005264285.1
12
1823
XM_006711971.2
XP_006712034.1
12
1823
XM_011532713.2
XP_011531015.1
12
1905
XM_011532714.2
XP_011531016.1
12
1754
XM_011532715.2
XP_011531017.1
12
1730
XM_011532716.2
XP_011531018.1
12
1823
XM_011532717.1
XP_011531019.1
12
1823
XM_011532718.2
XP_011531020.1
11
1484
XM_017003643.1
XP_016859132.1
12
1720
XM_017003644.1
XP_016859133.1
12
1697
XM_017003645.1
XP_016859134.1
12
1762
XM_017003646.1
XP_016859135.1
12
1825
XM_017003647.1
XP_016859136.1
11
1657
XM_017003648.1
XP_016859137.1
11
1653
XM_017003649.1
XP_016859138.1
11
1732
XM_017003650.1
XP_016859139.1
12
1762
XM_017003651.1
XP_016859140.1
11
1657
XM_017003652.1
XP_016859141.1
11
1718
XM_017003653.1
XP_016859142.1
11
1646
XM_017003654.1
XP_016859143.1
12
1823
XM_017003655.1
XP_016859144.1
11
1718
GenBank mRNA
AB020710.1
-
4
577
AK292545.1
-
11
1805
AL833968.1
-
12
1730
AY331186.1
-
12
1759
AY531390.1
-
10
1277
BC067215.1
-
11
1613
6502257
Back To Top
More Information
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-86c4h:80/10.239.166.160:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0