Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152760.2 | NP_689973.2 | 7 | 783 | ||
XM_006718488.3 | XP_006718551.1 | 7 | 1086 | |||
XM_017017470.1 | XP_016872959.1 | 7 | 1061 | |||
XM_017017471.1 | XP_016872960.1 | 8 | 1245 | |||
XM_017017472.1 | XP_016872961.1 | 6 | 1447 | |||
XM_017017473.1 | XP_016872962.1 | 6 | 1447 | |||
GenBank mRNA | AK055496.1 | - | 7 | 743 | ||
AK289587.1 | - | 7 | 749 | |||
AK296055.1 | - | 4 | 590 | |||
AK299512.1 | - | 6 | 853 | |||
AK309789.1 | - | 5 | 999 | |||
AL833261.1 | - | 7 | 769 | |||
BC040981.2 | - | 7 | 759 | 4797976 | ||
BC045563.1 | - | 7 | 732 | 4815203 |