Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZBTB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001123329.1 | NP_001116801.1 | 2 | 564 | ||
NM_014950.2 | NP_055765.2 | 2 | 564 | |||
XM_005267410.1 | XP_005267467.1 | 4 | 700 | |||
XM_011536566.2 | XP_011534868.1 | 3 | 437 | |||
XM_011536568.2 | XP_011534870.1 | 4 | 700 | |||
XM_017021094.1 | XP_016876583.1 | 4 | 1576 | |||
XM_017021095.1 | XP_016876584.1 | 3 | 492 | |||
GenBank mRNA | AB023214.1 | - | 2 | 435 | ||
AK291743.1 | - | 2 | 342 | |||
AY598321.1 | - | 1 | 173 | |||
BC050719.1 | - | 4 | 604 | 6141266 | ||
BX248777.1 | - | 2 | 564 |