Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LINC00935 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_029448.1 | 4 | 413 | |||
NR_029449.1 | 3 | 328 | ||||
GenBank mRNA | AK131005.1 | - | 3 | 328 | ||
BC036043.1 | - | 3 | 303 | |||
BC054859.1 | - | 4 | 395 | |||
BC146870.1 | - | 3 | 310 | |||
BC146877.1 | - | 3 | 310 |