Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC153 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145018.1 | NP_001138490.1 | 7 | 602 | ||
XM_006718818.3 | XP_006718881.1 | 4 | 482 | |||
XM_011542763.2 | XP_011541065.1 | 10 | 1089 | |||
XM_011542764.2 | XP_011541066.1 | 10 | 1110 | |||
XM_011542770.2 | XP_011541072.1 | 8 | 798 | |||
XM_011542771.2 | XP_011541073.1 | 6 | 768 | |||
XM_011542772.2 | XP_011541074.1 | 6 | 583 | |||
XM_011542774.2 | XP_011541076.1 | 9 | 933 | |||
XM_017017577.1 | XP_016873066.1 | 5 | 819 | |||
XM_017017579.1 | XP_016873068.1 | 5 | 367 | |||
XM_017017580.1 | XP_016873069.1 | 8 | 751 | |||
GenBank mRNA | BC043548.1 | - | 9 | 1674 | ||
BC101443.1 | - | 3 | 476 | 40022161 | ||
BC101444.1 | - | 3 | 476 | 40022159 | ||
BC101986.1 | - | 3 | 476 | 40022155 |