Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNRG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173605.1 | NP_775876.1 | 2 | 1014 | ||
NM_199464.2 | NP_955751.1 | 3 | 1112 | |||
GenBank mRNA | AK313401.1 | - | 3 | 963 | ||
AY129653.1 | - | 3 | 922 | |||
AY129654.1 | - | 2 | 832 | |||
AY169387.1 | - | 3 | 1112 | |||
AY169388.1 | - | 2 | 1014 | |||
AY190921.1 | - | 2 | 794 | |||
AY190922.1 | - | 3 | 892 | |||
BC020887.1 | - | 3 | 922 | 4690662 | ||
BC032343.1 | - | 2 | 808 | 5207702 |