Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM177A1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001079519.1 | NP_001072987.1 | 7 | 968 | ||
NM_001289022.1 | NP_001275951.1 | 6 | 691 | |||
NM_173607.3 | NP_775878.2 | 5 | 638 | |||
GenBank mRNA | AK096173.1 | - | 6 | 579 | ||
AL833508.1 | - | 5 | 552 | |||
BC029559.1 | - | 6 | 560 | 5310901 | ||
BX640884.1 | - | 5 | 641 | |||
BX640984.1 | - | 5 | 589 | |||
CR749796.1 | - | 5 | 638 |
Gene Symbol: LOC101927178 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110415.1 | 1 | 15 | |||
GenBank mRNA | CK904251.1 | - | 1 | 15 |