Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H3F3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005324.4 | NP_005315.1 | 4 | 993 | ||
GenBank mRNA | AF218029.1 | - | 4 | 817 | ||
AK130772.1 | - | 4 | 836 | |||
AK304607.1 | - | 4 | 766 | |||
BC001124.2 | - | 4 | 821 | 2988279 | ||
BC006497.2 | - | 4 | 821 | 2959756 | ||
BC012813.2 | - | 4 | 821 | 2959756 | ||
BC017558.1 | - | 4 | 825 | 3910031 | ||
BC020466.1 | - | 2 | 529 | |||
BC108701.1 | - | 4 | 800 | 6144255 | ||
BX537379.1 | - | 4 | 838 | |||
BX647280.1 | - | 4 | 836 |