Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POTEA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002920.1 | NP_001002920.1 | 4 | 663 | ||
NM_001005365.2 | NP_001005365.2 | 4 | 663 | |||
XM_011544516.1 | XP_011542818.1 | 4 | 735 | |||
GenBank mRNA | AY462869.1 | - | 4 | 663 | ||
AY462870.1 | - | 4 | 663 |